Learn More
Brady™ V4500 Barcode Scanner Battery Charger Power Supply Kit
Product Code. 30057746
Click to view available options
For Use With (Equipment):
V4500 Barcode Scanner (177233)
Unit Size:
1 set
Product Code. 30057746
Brand: Brady™ 177232
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
V4500 Barcode Scanner accessories to ensure uninterrupted productivity.
Battery Charger - V1300 Power Supply Kit for V4500 Barcode Scanner
Specifications
Description V1300 Kit for V4500 Barcode Reader
Height (Metric) 75 mm
Height (English) 2.95 in.
Width (Metric) 255 mm
Width (English) 10.04 in.
Depth (Metric) 113 mm
Depth (English) 4.45 in.
Weight (Metric) 0.79 kg
Charging Time Charging over battery charger (0 to 100%) 4 hr.
Weight (English) 1.742 lb.
Certifications/Compliance CE, UKCA
For Use With (Application) Asset Tracking, Barcode Labelling, Circuit Board Labelling, Component and Equipment Labelling, General Identification, Inventory and Inspection Labelling, Warehouse Marking
Humidity 5% to 95% (non-condensing)
For Use With (Equipment) V4500 Barcode Scanner (177233)
Includes (1) V1300 Cradle (177235), (1) V1200 Battery (177234), (1) AC Adapter (177241), (1) BRA-PWR-NA Power Cable (177242), (1) USB 1M Cable (177237)
Warranty 3 Years
Type Power Supply
Show More Show Less
Has lithium-ion battery - must ship ground