missing translation for 'onlineSavingsMsg'
Learn More

V-type proton ATPase subunit F Antibody, DyLight 488, Novus Biologicals™

Product Code. 18152970 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18152970 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18152970 Supplier Novus Biologicals Supplier No. NBP297190G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

V-type proton ATPase subunit F Polyclonal specifically detects V-type proton ATPase subunit F in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen V-type proton ATPase subunit F
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 488
Formulation 50 mM Sodium Borate
Gene Alias ATP6S14adenosinetriphosphatase 14k chain, ATPase, H+ transporting, lysosomal 14kDa, V1 subunit F, H(+)-transporting two-sector ATPase, 14kD subunit, MGC117321, MGC126037, MGC126038, vacuolar ATP synthase subunit F, vacuolar proton pump F subunit, Vacuolar proton pump subunit F, VATFATPase, vacuolar, 14 kD, V-ATPase 14 kDa subunit, V-ATPase F subunit, V-ATPase subunit F, Vma7, V-type proton ATPase subunit F
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human V-type proton ATPase subunit F fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 9296
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.