Learn More
UVP Mini Ultraviolet Lamp Replacement Tube
Product Code. 11796832
Change view
Click to view available options
Wavelength:
254/365 nm
365 nm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Wavelength unitSize
11796832 365 nm Each
11788231 254/365 nm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11796832 Supplier UVP Supplier No. 34001001
Only null left
Add to basket
Add to basket
Replacement tubes for lightweight, compact and financial UV Lamps
Specifications
Wattage 4 W
Wavelength 365 nm
For Use With (Equipment) Model UVL-4, UVL-23R Handheld UV Lamp, Mini UV Lamp
Packaging Quantity 1 Ea.