missing translation for 'onlineSavingsMsg'
Learn More

USP37 Antibody, Biotin, Novus Biologicals™

Product Code. 18551852 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18551852 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18551852 Supplier Novus Biologicals Supplier No. NB11040696B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

USP37 Polyclonal antibody specifically detects USP37 in Human samples. It is validated for Western Blot, Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen USP37
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Biotin
Dilution Western Blot, Immunohistochemistry
Formulation PBS
Gene Alias Deubiquitinating enzyme 37, EC 3.1.2.15, EC 3.4.19.12, KIAA1594tmp_locus_50, MGC117261, ubiquitin carboxyl-terminal hydrolase 37, ubiquitin specific peptidase 37, ubiquitin specific protease 37, ubiquitin thioesterase 37, Ubiquitin thiolesterase 37, Ubiquitin-specific-processing protease 37
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 125 and 175 of human Ubiquitin Specific Peptidase 37 using the numbering given in entry NP_065986.1 (GeneID 57695).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Turnover, Ubiquitin Proteasome Pathway
Primary or Secondary Primary
Gene ID (Entrez) 57695
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.