missing translation for 'onlineSavingsMsg'
Learn More

USP28 Antibody (S08-7I7), Novus Biologicals™

Product Code. 18649203 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
Unit Size:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18649203 100 μg 100µL
18617813 50 μg 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18649203 Supplier Novus Biologicals Supplier No. NBP319799100ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

USP28 Monoclonal antibody specifically detects USP28 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen USP28
Applications Western Blot
Classification Monoclonal
Clone S08-7I7
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Gene Alias Deubiquitinating enzyme 28, EC 3.1.2.15, EC 3.4.19.12, KIAA1515ubiquitin carboxyl-terminal hydrolase 28, ubiquitin specific peptidase 28, ubiquitin specific protease 28, ubiquitin thioesterase 28, Ubiquitin thiolesterase 28, Ubiquitin-specific-processing protease 28
Host Species Rabbit
Immunogen A synthetic peptide of human USP28 (Uniprot # Q96RU2)
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57646
Target Species Human
Content And Storage Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.