missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ USP21 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17147163
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17147163 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17147163 Supplier Invitrogen™ Supplier No. PA5109229

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive Control: Mouse thymus tissue lysate, PC-3M, SH-SY-5Y, human liver cancer tissue, human kidney tissue, human fetal skeletal muscle tissue.

This gene encodes a member of the C19 peptidase family, also known as family 2 of ubiquitin carboxy-terminal hydrolases. The encoded protein cleaves ubiquitin from ubiquitinated proteins for recycling in intracellular protein degradation. The encoded protein is also able to release NEDD8, a ubiquitin-like protein, from NEDD8-conjugated proteins. This gene has been referred to as USP16 and USP23 but is now known as USP21. Alternatively spliced transcript variants have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen USP21
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.2% BSA and 0.05% sodium azide; pH 7.4
Gene USP21
Gene Accession No. Q9UK80
Gene Alias deubiquitinating enzyme 21; ESTM28; NEDD8-specific protease; PP1490; similar to Ubiquitin carboxyl-terminal hydrolase 21 (Ubiquitin thiolesterase 21) (Ubiquitin-specific processing protease 21) (Deubiquitinating enzyme 21) (NEDD8-specific protease); ubiquitin carboxyl-terminal hydrolase 21; ubiquitin specific peptidase 21; ubiquitin specific protease 21; ubiquitin specific protease 23; ubiquitin thioesterase 21; ubiquitin thiolesterase 21; ubiquitin-specific processing protease 21; ubiquitin-specific protease 16; ubiquitin-specific protease 21; ubiquitin-specific-processing protease 21; USP16; USP21; Usp23; W53272
Gene Symbols USP21
Host Species Rabbit
Immunogen Synthetic peptide within Human USP21 aa 1-50.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27005
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.