Learn More
Uridine 5’-Monophosphate Disodium Salt, TRC
Product Code. 17965075
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17965075 1 g 1g
30136673 10 g 10g
30136199 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17965075 Supplier Toronto Research Chemicals Supplier No. TRCU8300051G
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3387-36-8
Formula Weight 367.9998
InChI Formula InChI=1S/C9H13N2O9P.2Na/c12-5-1-2-11(9(15)10-5)8-7(14)6(13)4(20-8)3-19-21(16,17)18;;/h1-2,4,6-8,13-14H,3H2,(H,10,12,15)(H2,16,17,18);;/q;2*+1/p-2/t4-,6-,7-,8-;;/m1../s1
IUPAC Name disodium;[(2R,3S,4R,5R)-5-(2,4-dioxopyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate
Molecular Formula C9 H11 N2 O9 P . 2 Na
Molecular Weight (g/mol) 368.14
SMILES [Na+].[Na+].O[C@H]1[C@@H](O)[C@@H](O[C@@H]1COP(=O)([O-])[O-])N2C=CC(=O)NC2=O
Synonym Uridine 5'-Monophosphate Disodium, 5'-Uridylic acid, disodium salt (7CI,8CI,9CI), 5'-UMP disodium salt, Disodium 5'-UMP, Disodium 5'-uridylate, Disodium UMP, Disodium uridine 5'-monophosphate, Disodium uridine 5'-phosphate, Disodium uridine monophosphate, NSC 20257, UMP disodium salt, Uridine 5'-(dihydrogen phosphate) disodium salt, Uridine 5'-monophosphate disodium salt, Uridylic acid disodium salt, 5'-Uridylic acid, sodium salt (1:2)
Chemical Name or Material Uridine-5'-monophosphate disodium salt
Specifications
Quantity 1 g