missing translation for 'onlineSavingsMsg'
Learn More

Uridine-5'-diphosphate trisodium salt, 98+%, Thermo Scientific Chemicals

Product Code. p-7041759
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15402969 100 mg 100mg
15412969 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15402969 Supplier Thermo Scientific Alfa Aesar Supplier No. J60141.MC
Add to basket
This item is not returnable. View return policy

It has been used as a purinergic agonist to study its effects on absorptive cationic flux in cochlear outer sulcus cells (OSC) and vestibular transitional cells (VTC). Also been used for nucleoside diphosphatase (NDPase) activity assays in rabbit retinae.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It has been used as a purinergic agonist to study its effects on absorptive cationic flux in cochlear outer sulcus cells (OSC) and vestibular transitional cells (VTC). Also been used for nucleoside diphosphatase (NDPase) activity assays in rabbit retinae.

Solubility
Soluble in water to 100mM

Notes
Store in freezer -20°C. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21931-53-3
Molecular Formula C9H11N2Na3O12P2
Molecular Weight (g/mol) 470.106
MDL Number MFCD00006524
InChI Key GFASEGWJDXDCKZ-PRYCXDIZSA-K
Synonym uridine 5'-diphosphate sodium salt, uridine 5'-diphosphate sodium salt from
PubChem CID 126968597
IUPAC Name trisodium;[[(2S,3S,4R,5R)-5-(2,4-dioxopyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-oxidophosphoryl] phosphate
SMILES C1=CN(C(=O)NC1=O)C2C(C(C(O2)COP(=O)([O-])OP(=O)([O-])[O-])O)O.[Na+].[Na+].[Na+]

Specifications

Color White
Quantity 100 mg
Solubility Information Soluble in water to 100mM
Formula Weight 470.11
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material Uridine-5'-diphosphate trisodium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.