Learn More
Urethane-d5, TRC
Specifications
Specifications
| Chemical Name or Material | Urethane-d5 |
| CAS | 73962-07-9 |
| Molecular Formula | C3 2H5 H2 N O2 |
| Quantity | 50 mg |
| Synonym | Ethan-d5-ol','carbamate (9CI), Carbamic acid ethyl-d5 ester, Ethyl-d5 carbamate, Urethane-d5, Urethane-D5 (ethyl-D5) |
| InChI Formula | InChI=1S/C3H7NO2/c1-2-6-3(4)5/h2H2,1H3,(H2,4,5)/i1D3,2D2 |
| SMILES | [2H]C([2H])([2H])C([2H])([2H])OC(=O)N |
| IUPAC Name | 1,1,2,2,2-pentadeuterioethyl carbamate |
| Molecular Weight (g/mol) | 94.12 |
| Formula Weight | 94.0791 |