Learn More
Thermo Scientific™ Uracil-DNA Glycosylase, heat-labile
Description
Uracil-DNA Glycosylase excises uracil from dU-containing DNA by cleaving the N-glycosidic bond between the uracil base and the sugar backbone. This cleavage generates alkali sensitive apyrimidinic sites that are blocked from replication by DNA polymerase or prevented from becoming a hybridization site.
Double and single-stranded dU-containing DNA are substrates for Uracil-DNA Glycosylase whereas RNA and normal dT-containing DNA are not.
Uracil-DNA Glycosylase can also be used to increase the cloning efficiency of PCR products having dU-containing primers incorporated into them as well as increasing the efficiency of site-directed mutagenesis.
The Uracil-DNA Glycosylase derived from Gadus morhua has all the attributes of the enzyme derived from E. coli with the added benefit of being heat-labile. It is completely and irreversibly inactivated after 10 minute incubation at 50°C.
Purity:
The enzyme is chromatographically purified and tested for contaminating endonucleases and exonucleases.
Storage Buffer:
20 mM Tris-HCl pH 7.5 (25°C), 50 mM NaCl, 0.4% non-ionic surfactant, 7 mM DTT, 0.1 mM EDTA, 50% Glycerol.
Assay Conditions:
The reaction mixture contains 70 mM Tris-HCl, pH 8.0, 10 mM NaCl, 1 mM EDTA, 100 μg/mL BSA, 3H-dUTP labeled DNA, and Uracil-DNA Glycosylase. Incubation is at 37°C for 1 hour.
Unit Definition:
One unit is the amount of enzyme required to liberate 1 nmol uracil from dU-containing DNA in one hour at 37°C.
Concentration:
1 unit/μL
Applications:
- Study of DNA repair and mutation detection.
- Increase cloning efficiency of PCR products.
- Increase the efficiency of site-directed mutagenesis.
- Study of protein-DNA interactions.
Source:
Recombinant from Gadus morhua (cod liver)
Specifications
Specifications
| Content And Storage | Shipped on dry ice. Store at -20°C. |
| Enzyme | Glycosylase |
| Compatible Buffer | Storage Buffer |
| Quantity | 100 U |
| Product Type | Uracil DNA Glycosylase |