Learn More
Uracil-6-d1, CDN
Product Code. 30322293
Change view
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30322293 0.05 g 0.05g
30324548 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30322293 Supplier CDN Supplier No. CDND51330.05G
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Uracil-6-D
CAS 24897-51-6
Molecular Formula C4 D H3 N2 O2
Quantity 0.05 g
Synonym 2,4(1 H,3 H)-Pyrimidinedione-6-d (9 CI, ACI), Uracil-6-d (7 CI, 8 CI), Uracil-6-D
InChI Formula InChI=1 S/C4H4N2O2/c7-3-1-2-5-4(8)6-3/h1-2 H,(H2,5,6,7,8)/i2D
SMILES [2 H]C1=CC(=O)NC(=O)N1
IUPAC Name 6-deuterio-1 H-pyrimidine-2,4-dione
Molecular Weight (g/mol) 113.093
Formula Weight 113.034 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity