missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GFL Gesellschaft Fuer Labortec™ Upright Freezer
Click to view available options
Capacity (Metric):
500 L
Unit Size:
Each
Specifications
Specifications
| Type | Freezer |
| Capacity (Metric) | 500 L |
| Temperature Range | -50°C to -85°C |
| Inner Doors | Yes |
| Style | Upright |
| For Use With (Equipment) | Only a reliable cooling technology and a first-class insulation ensure trouble-free long-term storage of cell lines, human, animal and plant tissues, fractions of the blood, vaccines, or reference cultures of micro-organisms, such as bacteria, yeasts, vir |
| Model | 6485 |
| Dimensions (D x W x H) Interior | 760 x 600 x 1100 mm |
| Depth (Metric) Exterior | 1095 mm |
| Height (Metric) Exterior | 1940 mm |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction