Learn More
Benchmark Scientific™ Universal Magnetic Platform for Flasks and Tube Racks
Description
- Magnetic platform holds magnetic clamps in place
- 16 x 16.5 in., fit Fisherbrand™ IncuShaker 10 L
- Clamps sold separately
Specifications
Specifications
| Holds | Magnetic or MAGic Clamps |
| Tube Capacity | Dependent upon Size and Rack |
| Clamps | Works with Magnetic or MAGic Clamps |
| Model | BT4010-MR |
| For Use With (Application) | Magnetic or MAGic Clamps |
| For Use With (Equipment) | Fisherbrand IncuShaker 10 L |