Learn More
Benchmark Scientific™ Universal Magnetic Platform for Flasks and Tube Racks
Product Code. 17994392
Change view
Click to view available options
Holds:
Magnetic or MAGic Clamps
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds unitSize
17994392 Magnetic or MAGic Clamps Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17994392 Supplier Benchmark Scientific™ Supplier No. BT4010MR
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
Universal Magnetic Platform for flasks & tube racks accommodates magnetic clamps in the FisherBrand™ IncuShaker 10 L.
  • Magnetic platform holds magnetic clamps in place
  • 16 x 16.5 in., fit Fisherbrand™ IncuShaker 10 L
  • Clamps sold separately
Specifications
Holds Magnetic or MAGic Clamps
Tube Capacity Dependent upon Size and Rack
Clamps Works with Magnetic or MAGic Clamps
Model BT4010-MR
For Use With (Application) Magnetic or MAGic Clamps
For Use With (Equipment) Fisherbrand IncuShaker 10 L