Learn More
Ohaus™ Universal Holder Cover
Product Code. 15772639 Shop All Ohaus Products
Click to view available options
For Use With (Equipment):
Universal Holder
Unit Size:
Each
Product Code. 15772639
Brand: Ohaus™ 30400225
Only null left
Add to basket
Add to basket
OHAUS™ Universal Holder Cover suitable for use with heavy duty vortex mixers.
  • OHAUS offers a large selection of accessories that can be used on all OHAUS vortex mixers
  • Increase throughput by mixing multiple samples at one time
  • Effortlessly mix up to 52 microtubes or eight microplates
  • Choose the appropriate holder for the sample size, or select the flat insert and cut out to suit custom specifications
TRUSTED_SUSTAINABILITY
Specifications
Product Type Cover
Color Black
Description Cover For Universal Holder
For Use With (Equipment) Universal Holder