Learn More
Poulten & Graf GmbH Universal Drain Tube
Description
- Flexible remote dispensing
- Tubing and dispensing valve are made of PTFE
Specifications
Specifications
| Description | Remote dispensing for OPTIFIX dispensers |
| Type | Spare part for OPTIFIX SAFETY, SAFETY S and OPTIMAT 3 |
| Volume (Metric) | 30 to 50 mL |
| For Use With (Equipment) | OPTIFIX SAFETY, SAFETY S and OPTIMAT 3 |
| Quantity | 1 |
| Electrical Requirements | No |