Learn More
Poulten & Graf GmbH Universal Drain Tube
Product Code. 10303763
Change view
Click to view available options
:
Volume Metric Range: 30 - 50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10303763 Volume Metric Range: 30 - 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10303763 Supplier Poulten & Graf GmbH Supplier No. 101 10402
Please to purchase this item. Need a web account? Register with us today!
Poulten & Graf GmbH Universal Drain Tube is a flexible tube that it compatable with OPTIFIX™ SAFETY, OFTIFIX™ SAFETY S, and OPTIMAT™ 3.
  • Flexible remote dispensing
  • Tubing and dispensing valve are made of PTFE
Specifications
Description Remote dispensing for OPTIFIX dispensers
Type Spare part for OPTIFIX SAFETY, SAFETY S and OPTIMAT 3
Volume (Metric) 30 to 50 mL
For Use With (Equipment) OPTIFIX SAFETY, SAFETY S and OPTIMAT 3
Quantity 1
Electrical Requirements No