Learn More
Sartorius UNF 10-32 Female to Luer Female
Product Code. 17879366 Shop All Sartorius Products
Click to view available options
Packaging Quantity:
2
Unit Size:
1 set
Product Code. 17879366
Brand: Sartorius 1ZA0005
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Adapter UNF 10-32 female to Luer female for Sartobind™ MA units.
  • Adapters to convert the Luer outlet connector on any Sartobind™ Lab unit to the UNF 10-32 standard. This enables the operation of Sartobind™ Lab units on FPLC systems fitted with UNF 10-32 male connectors.
  • This is a replacement part - a minimum of one piece is already provided in each box of Sartobind™ Lab unit(s).
  • Note: One Luer male/UNF 10-32 female adapter is also required, to adapt the inlet connector on Sartobind™ Lab units to the UNF 10-32 standard. A minimum of one piece is already provided in each box of Sartobind™ Lab unit(s), and replacements are available to purchase separately
  • Adapters for connecting Sartobind™ Lab units to most liquid chromatography systems
  • Easily convert female Luer lock inlet connectors to the UNF 10-32 standard
  • Tool-free, fingertight connection
TRUSTED_SUSTAINABILITY
Specifications
Type Adapter
Packaging Quantity 2
Material PEEK
Pressure 100 psi