Learn More
Sartorius UNF 10-32 Female to Luer Female
Shop All Sartorius Products
Click to view available options
Packaging Quantity:
2
Unit Size:
1 set
Description
- Adapters to convert the Luer outlet connector on any Sartobind™ Lab unit to the UNF 10-32 standard. This enables the operation of Sartobind™ Lab units on FPLC systems fitted with UNF 10-32 male connectors.
- This is a replacement part - a minimum of one piece is already provided in each box of Sartobind™ Lab unit(s).
- Note: One Luer male/UNF 10-32 female adapter is also required, to adapt the inlet connector on Sartobind™ Lab units to the UNF 10-32 standard. A minimum of one piece is already provided in each box of Sartobind™ Lab unit(s), and replacements are available to purchase separately
- Adapters for connecting Sartobind™ Lab units to most liquid chromatography systems
- Easily convert female Luer lock inlet connectors to the UNF 10-32 standard
- Tool-free, fingertight connection
Specifications
Specifications
| Type | Adapter |
| Packaging Quantity | 2 |
| Material | PEEK |
| Pressure | 100 psi |