missing translation for 'onlineSavingsMsg'
Learn More

Undecanophenone, 97%, Thermo Scientific™

Product Code. p-7043691
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11311517

Brand: Thermo Scientific Alfa Aesar L02119.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4433-30-1
Molecular Formula C17H26O
Molecular Weight (g/mol) 246.394
MDL Number MFCD00051548
InChI Key LHJBFOGCFZHBAJ-UHFFFAOYSA-N
Synonym undecanophenone, n-undecanophenone, 1-phenylundecanone, 1-undecanone, 1-phenyl, decyl phenyl ketone, 1-phenyl-1-undecanone, 1-phenyl-undecan-1-one, 1-phenyl-1-undecanone #, acmc-209jx6
PubChem CID 78167
IUPAC Name 1-phenylundecan-1-one
SMILES CCCCCCCCCCC(=O)C1=CC=CC=C1

Specifications

Melting Point 28°C to 30°C
Boiling Point 202°C to 204°C (14 mmHg)
Quantity 5 g
Beilstein 2211502
Formula Weight 246.39
Percent Purity 97%
Chemical Name or Material Undecanophenone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.