missing translation for 'onlineSavingsMsg'
Learn More

UNC13D/Munc 13-4 Antibody (1224B) [mFluor Violet 500 SE], Novus Biologicals™

Product Code. 30678796 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30678796 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30678796 Supplier Novus Biologicals Supplier No. FAB89662MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

UNC13D/Munc 13-4 Monoclonal antibody specifically detects UNC13D/Munc 13-4 in Human,Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen UNC13D/Munc 13-4
Applications Western Blot
Classification Monoclonal
Clone 1224B
Conjugate mFluor Violet 500 SE
Formulation 50mM Sodium Borate
Gene Alias FHL3, HLH3, HPLH3, MUNC13-4, Munc13-4protein unc-13 homolog D, unc-13 homolog D (C. elegans)
Host Species Rabbit
Immunogen KLH-coupled N-terminal human UNC13D peptide, Accession # Q70J99
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 201294
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.