Learn More
Tocris Bioscience™ UNC 1999
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Description
UNC 1999 is a potent and SAM-competitive EZH2/EZH1 lysine methyltransferase inhibitor (IC50 values are 10 and 45 nM, respectively). Selective for EZH2/EZH1 over a panel of other methyltransferases and non-epigenetic targets. Reduces H3K27me3 levels in vitro. Prolongs survival of MLL-AF9 bearing mice. Orally bioavailable.
Negative Control also available.
Specifications
Specifications
| Quantity | 50 mg |
| Product Type | UNC 1999 |
| Purity | >99% |
| Molecular Weight (g/mol) | 569.74 |
| Inhibitors | UNC 1999 |