missing translation for 'onlineSavingsMsg'
Learn More

UNC 0631, MedChemExpress

Product Code. p-200101387
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16492017 50 mg 50mg
16485868 10 mg 10mg
16473386 25 mg 25mg
16408527 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16492017 Supplier Medchem Express Supplier No. HY13808

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress UNC 0631 is a potent histone methyltransferase G9a inhibitor with an IC50 of 4 nM. UNC 0631 potently reduces H3K9me2 levels in MDA-MB-231 cells with an IC50 of 25 nM.

UNC 0631 is a potent histone methyltransferase G9a inhibitor with an IC50 of 4 nM. UNC 0631 potently reduces H3K9me2 levels in MDA-MB-231 cells with an IC50 of 25 nM.

Specifications

Chemical Name or Material UNC 0631
CAS 1320288-19-4
Color Yellow
Purity Grade Notes Research
Molecular Formula C37H61N7O2
Quantity 50 mg
Solubility Information DMF : 150 mg/mL (235.88 mM; Need ultrasonic) ∣Ethanol : 100 mg/mL (157.25 mM; Need ultrasonic) ∣DMSO : 16.67 mg/mL (26.21 mM; Need ultrasonic)
SMILES COC1=CC2=C(NC3CCN(CC4CCCCC4)CC3)N=C(N5CCN(C(C)C)CCC5)N=C2C=C1OCCCN6CCCCC6
Molecular Weight (g/mol) 635.93
Formula Weight 635.93
Percent Purity 99.35%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.