Learn More
WTW™ Ultrapure Water Conductivity Cell
Product Code. 10387073
Click to view available options
For Use With (Equipment):
Integrated Temperature Sensor
Unit Size:
Each
This item is not returnable. View return policy
Product Code. 10387073
Brand: WTW™ 301992
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is not returnable. View return policy
Includes flow vessels to prevent wrong results due to by carbon dioxide
Pure water measurement means basically the intrinsic conductivity of purified water in the pharmaceutical industry and the production of semiconductors. This is where the LR 325/01 and the LR 325/001 come in. Both have flow vessels to prevent carbon dioxide from the air entering during the measurement process. The LR 01/V is the same as the LR 325/01 and can be connected to the Vario Cond. The LR 325/001 is capable of measuring into ranges of 0.001 µS/cm when using the suitable WTW conducitivity meters.
  • Precise measured values thanks to concentric electrodes without edge fields
  • Includes flow vessels to prevent wrong results due to by carbon dioxide
  • No polarization effects thanks to stainless steel design
TRUSTED_SUSTAINABILITY
Specifications
Type Ultrapure Water Conductivity Cell
For Use With (Equipment) Integrated Temperature Sensor