Beschreibung
- Ideal as over-glasses for conventional prescription glasses
- Ultralight for maximum wearing comfort
- Safety spectacles without hinges, completely metal-free
- Optimum optical clarity
Spezifikation
Spezifikation
| Certifications/Compliance | DIN EN 166 - 168 and 170 |
| Lens Coating | Scratch-resistant |
| Lens Tint | Clear |
| Product Type | Over-the-Glasses Safety Glasses |
| For Use With (Application) | Eye Protection |
| Frame Color | Blue |
| No. per Pack | 1 Ea. |
| Material | Polycarbonate |
| Disposable | Limited-use |
| Lens Material | Polycarbonate |
| Mehr anzeigen |