missing translation for 'onlineSavingsMsg'
Learn More

ULBP-4/RAET1E Antibody (709116) [Janelia Fluor™ 669], Novus Biologicals™

Product Code. 30164222 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30164222 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30164222 Supplier Novus Biologicals Supplier No. FAB6285JF669

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ULBP-4/RAET1E Monoclonal antibody specifically detects ULBP-4/RAET1E in Human samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen ULBP-4/RAET1E
Applications Flow Cytometry, Immunohistochemistry, Western Blot, CyTOF
Classification Monoclonal
Clone 709116
Conjugate Janelia Fluor 669
Formulation 50mM Sodium Borate
Gene Alias bA350J20.7, LETALRAE-1-like transcript 4, Lymphocyte effector toxicity activation ligand, MGC125309, N2DL4, N2DL-4, NKG2D ligand 4, NKG2DL4, RAET1E2, retinoic acid early transcript 1ERL-4, ULBP4MGC125308
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human ULBP-4/RAET1E, Gly30-Asp225, Accession # Q8TD07
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 135250
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.