missing translation for 'onlineSavingsMsg'
Learn More

UEVLD Monoclonal Antibody (OTI8G3), TrueMAB™, OriGene

Product Code. 17571204
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17571204 30 μL 30µL
17551204 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17571204 Supplier Origene Technologies Supplier No. TA809206S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

UEVLD Monoclonal antibody specifically detects UEVLD in Human samples. It is validated for Western Blot

Specifications

Antigen UEVLD
Applications Western Blot
Classification Monoclonal
Clone OTI8G3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Ncf4
Gene Accession No. Q15080
Gene Alias 8430408E05Rik; Attp; EV and lactate/malate dehydrogenase domain-containing protein; signaling molecule ATTP; ubiquitin-conjugating enzyme E2 variant 3; UEV and lactate/malate dehyrogenase domains; Uev3; UEV-3; UEVLD
Host Species Mouse
Immunogen Full length human recombinant protein of human NCF4 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4689
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.