missing translation for 'onlineSavingsMsg'
Learn More

UCK2 Antibody (OTI21A6) [Biotin], Novus Biologicals™

Product Code. 30682721 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30682721 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30682721 Supplier Novus Biologicals Supplier No. NBP344951B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

UCK2 Monoclonal antibody specifically detects UCK2 in Human,Mouse,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen UCK2
Applications Western Blot
Classification Monoclonal
Clone OTI21A6
Conjugate Biotin
Formulation PBS
Gene Alias Cytidine monophosphokinase 2, DKFZp686M04245, EC 2.7.1.48, Testis-specific protein TSA903, TSA903, UCK 2, UK, UMPK, uridine kinase, uridine monophosphate kinase, Uridine monophosphokinase 2, uridine-cytidine kinase 2
Host Species Mouse
Immunogen Synthetic peptide corresponding to residues near C-terminus of human UCK2 (NP_036606).
Purification Method >95%
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 7371
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.