missing translation for 'onlineSavingsMsg'
Learn More

UC-112, MedChemExpress

Product Code. 16415398
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16415398

Brand: Medchem Express HY12842

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress UC-112 is a novel potent IAP(Inhibitor of apoptosis) inhibitor; potently inhibit cell growth in two human melanoma (A375 and M14) and two human prostate (PC-3 and DU145) cancer cell lines(IC50=0.7-3.4 uM).

UC-112 is a novel potent IAP(Inhibitor of apoptosis) inhibitor

Specifications

Chemical Name or Material UC-112
CAS 383392-66-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C22H24N2O2
Quantity 10 mg
Solubility Information DMSO : 33.33 mg/mL (95.65 mM; Need ultrasonic)
SMILES OC1=C2N=CC=CC2=C(COCC3=CC=CC=C3)C=C1CN4CCCC4
Molecular Weight (g/mol) 348.44
Formula Weight 348.44
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.