Learn More
UC-112, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
UC-112 is a novel potent IAP(Inhibitor of apoptosis) inhibitor
Specifications
Specifications
| Chemical Name or Material | UC-112 |
| CAS | 383392-66-3 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C22H24N2O2 |
| Quantity | 50 mg |
| Solubility Information | DMSO : 33.33 mg/mL (95.65 mM; Need ultrasonic) |
| SMILES | OC1=C2N=CC=CC2=C(COCC3=CC=CC=C3)C=C1CN4CCCC4 |
| Molecular Weight (g/mol) | 348.44 |
| Formula Weight | 348.44 |
| Show More |