missing translation for 'onlineSavingsMsg'
Learn More

UBXN2A Antibody, HRP, Novus Biologicals™

Product Code. 18688062 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18688062 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18688062 Supplier Novus Biologicals Supplier No. NBP298564H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

UBXN2A Polyclonal antibody specifically detects UBXN2A in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBXN2A
Applications Western Blot
Classification Polyclonal
Conjugate HRP
Formulation PBS
Gene Alias MGC138202, UBX domain containing 4, UBX domain protein 2A, UBX domain-containing protein 2A, UBX domain-containing protein 4, UBXD4
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human UBXN2A fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 165324
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.