Learn More
UBR5 Sheep anti-Human, Mouse, Rat, Polyclonal, R&D Systems™
GREENER_CHOICE
Sheep Polyclonal Antibody
Brand:  R&D Systems AF6404-SP
Encompass
Product Code. 15709142
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
This item is not returnable. View return policy
Description
Description
UBR5/EDD Polyclonal specifically detects UBR5/EDD in Human, Mouse, Rat samples. It is validated for Western Blot.