missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ UBP 310

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
UBP 310 is a GluK1 (formerly GLUK5) kainate receptor antagonist (IC50 = 130 nM); also blocks recombinant homomeric GluK3 (forrmerly GLUK7) receptors. Displays 12,700-fold selectivity for GluK1 (formerly GLUK5) over GluK2 (formerly GLUK6). Exhibits no activity at mGlu group I or NMDA receptors at concentrations of up to 10 μM. Apparent KD value is 18 ± 4 nM for depression of kainate responses on the dorsal root.
Please refer to IUPHAR Guide to Pharmacology for the most recent naming conventions.
Specifications
Specifications
| Quantity | 10 mg |
| Product Type | UBP 310 |
| Purity | >98% |
| Molecular Weight (g/mol) | 353.35 |
| Inhibitors | UBP 310 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction