missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ubiquilin 2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 6 publications
314.00 € - 542.00 €
Specifications
| Antigen | Ubiquilin 2 |
|---|---|
| Dilution | Western Blot 0.4 ug/ml, Simple Western 1:20, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50-1:200 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18454522
|
Novus Biologicals
NBP1-85639-25ul |
25 μL |
314.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18777433
|
Novus Biologicals
NBP1-85639 |
542.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
Ubiquilin 2 Polyclonal specifically detects Ubiquilin 2 in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
| Ubiquilin 2 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 29978 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:LSAMSNPRAMQALMQIQQGLQTLATEAPGLIPSFTPGVGVGVLGTAIGPVGPVTPIGPIGPIVPFTPIGPIGPIGPTGPAAPPGSTGSGGPTGPTVSSAAPSETTSPTSESGPNQQFIQQMVQALAGANAPQLPNPEVRFQQQLEQLN | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot 0.4 ug/ml, Simple Western 1:20, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50-1:200 | |
| Polyclonal | |
| Rabbit | |
| Human, Mouse | |
| Chap1, CHAP1/DSK2, DSK2, DSK2 homolog, FLJ10167, FLJ56541, hPLIC-2, N4BP4LIC-2, Nedd4 binding protein 4, PLIC-2, PLIC2Dsk2, Protein linking IAP with cytoskeleton 2, RIHFB2157, ubiquilin 2, ubiquilin-2, Ubiquitin-like product Chap1/Dsk2 | |
| UBQLN2 | |
| IgG | |
| Affinity Purified | |
| Specificity of human Ubiquilin 2 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title