missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UbcH5c/UBE2D3 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
188.00 € - 470.00 €
Specifications
| Antigen | UbcH5c/UBE2D3 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18627150
|
Novus Biologicals
NBP2-93898-0.02ml |
0.02 mL |
188.00 €
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18663431
|
Novus Biologicals
NBP2-93898-0.1ml |
0.1 mL |
470.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
UbcH5c/UBE2D3 Polyclonal antibody specifically detects UbcH5c/UBE2D3 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| UbcH5c/UBE2D3 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Biologically Active Proteins | |
| PBS (pH 7.3), 50% glycerol | |
| 7323 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| E2(17)KB 3, E2(17)KB3, EC 6.3.2.19, MGC43926, MGC5416, UBC4/5, UBC5C, UBCH5C, Ubiquitin carrier protein D3, ubiquitin-conjugating enzyme E2 D3, Ubiquitin-conjugating enzyme E2(17)KB 3, Ubiquitin-conjugating enzyme E2-17 kDa 3, ubiquitin-conjugating enzyme E2D 3 (homologous to yeast UBC4/5), ubiquitin-conjugating enzyme E2D 3 (UBC4/5 homolog, yeast), Ubiquitin-protein ligase D3 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human UBE2D3 (NP_003331.1). MALKRINKELSDLARDPPAQCSAGPVGDDMFHWQATIMGPNDSPYQGGVFFLTIHFPTDYPFKPPKVAFTTRIYHPNINSNGSICLDILRSQWSPALTIS | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title