Learn More
SI Analytics UBBELOHDE TUBE 0,3 A 1mm2/S
Brand:  SI Analytics 285400164
UBBELOHDE TUBE 0,3 A 1mm2/S
Product Code. 10004110
  • ONLINE EXCLUSIVE PRICE
    537.00 €
    478.00 € / Each
    Save 59.00 €
    10% Off
Only null left
Estimated Shipment: 05-02-2026