Learn More
U93631, MedChemExpress
Product Code. 16495847
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16495847 5 mg 5mg
16473118 10 mg 10mg
16439676 50 mg 50mg
16414416 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16495847 Supplier Medchem Express Supplier No. HY100686
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
MedChemExpress U93631 is a GABAA receptor ligand of novel chemical structure with IC50 of 100 nM,and has been shown to induce a rapid, time-dependent decay of GABA-induced whole-cell Cl-currents in recombinant GABAA receptors.
U93631 is a GABAA receptor ligand of novel chemical structure with IC50 of 100 nM,and has been shown to induce a rapid, time-dependent decay of GABA-induced whole-cell Cl-currents in recombinant GABAA receptors.
Specifications
Chemical Name or Material U93631
CAS 152273-12-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C17H21N3O2
Quantity 5 mg
Solubility Information DMSO : ≥ 34 mg/mL (113.57 mM)
SMILES O=C(C1=C2N(C=N1)C3=C(C=CC=C3)NC2(C)C)OC(C)(C)C
Molecular Weight (g/mol) 299.37
Formula Weight 299.37
Percent Purity 99.85%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation