missing translation for 'onlineSavingsMsg'
Learn More

U0126-EtOH, MedChemExpress

Product Code. 16461026
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
This item is not returnable. View return policy

Product Code. 16461026

Brand: Medchem Express HY12031

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress U0126 (U0126-EtOH) is a potent, non-ATP competitive and selective MEK1 and MEK2 inhibitor, with IC50s of 72 nM and 58 nM, respectively. U0126 is an autophagy and mitophagy inhibitor.

U0126 (U0126-EtOH) is a potent, non-ATP competitive and selective MEK1 and MEK2 inhibitor, with IC50s of 72 nM and 58 nM, respectively. U0126 is an autophagy and mitophagy inhibitor.

Specifications

Chemical Name or Material U0126-EtOH
Physical Form Solid
CAS 1173097-76-1
Color Off-White
Quantity 200 mg
Purity Grade Notes Research
Molecular Formula C20H22N6OS2
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : 50 mg/mL (117.22 mM; Need ultrasonic)
SMILES NC1=CC=CC=C1S/C(N)=C(/C(C#N)=C(SC2=C(C=CC=C2)N)\N)C#N.CCO
Molecular Weight (g/mol) 426.56
Formula Weight 426.56
Percent Purity 96.54%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.