missing translation for 'onlineSavingsMsg'
Learn More

Tyrphostin AG 528, MedChemExpress

Product Code. 16463048
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16463048

Brand: Medchem Express HY100499

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Tyrphostin AG 528 is an inhibitor of EGFR and ErbB2 with IC50s of 4.9 and 2.1 μM, respectively. Tyrphostin AG 528 (Tyrphostin B66) is a protein tyrosine kinase inhibitor, with IC50s of 4.9 μM for epidermal growth factor receptors (EGFR) and 2.1 μM for ErbB2. Tyrphostin AG 528 is also an anticancer agent.

Tyrphostin AG 528 is an inhibitor of EGFR and ErbB2 with IC50s of 4.9 and 2.1 μM, respectively.

Specifications

Chemical Name or Material Tyrphostin AG 528
CAS 133550-49-9
Color Yellow
Purity Grade Notes Research
Molecular Formula C18H14N2O3
Quantity 10 mg
Synonym Tyrphostin B66 AG 528
Solubility Information DMSO : 25 mg/mL (81.61 mM; Need ultrasonic)
SMILES O=C(/C(C#N)=C/C1=CC=C(O)C(O)=C1)N2CCC3=CC=CC=C32
Molecular Weight (g/mol) 306.32
Formula Weight 306.32
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.