missing translation for 'onlineSavingsMsg'
Learn More

Tyrphostin A1, MedChemExpress

Product Code. p-200101534
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16487136 100 mg 100mg
16417118 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16487136 Supplier Medchem Express Supplier No. HY16668

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Tyrphostin A1(AG9) inhibits CD40L-stimulated IL-12 production in macrophage cultures and antigen-induced generation of Th1 cells.

Tyrphostin A1(AG9) inhibits CD40L-stimulated IL-12 production in macrophage cultures and antigen-induced generation of Th1 cells.

Specifications

Chemical Name or Material Tyrphostin A1
CAS 2826-26-8
Color Yellow
Purity Grade Notes Research
Molecular Formula C11H8N2O
Quantity 100 mg
Synonym Tyrphostin 1 AG9
Solubility Information DMSO : ≥ 100 mg/mL (542.92 mM)
SMILES N#C/C(C#N)=C\C1=CC=C(OC)C=C1
Molecular Weight (g/mol) 184.19
Formula Weight 184.19
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.