Learn More
Tyrphostin A1, MedChemExpress
Product Code. 16487136
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16487136 100 mg 100mg
16417118 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16487136 Supplier Medchem Express Supplier No. HY16668
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Tyrphostin A1(AG9) inhibits CD40L-stimulated IL-12 production in macrophage cultures and antigen-induced generation of Th1 cells.
Tyrphostin A1(AG9) inhibits CD40L-stimulated IL-12 production in macrophage cultures and antigen-induced generation of Th1 cells.
Specifications
Chemical Name or Material Tyrphostin A1
CAS 2826-26-8
Color Yellow
Purity Grade Notes Research
Molecular Formula C11H8N2O
Quantity 100 mg
Synonym Tyrphostin 1 AG9
Solubility Information DMSO : ≥ 100 mg/mL (542.92 mM)
SMILES N#C/C(C#N)=C\C1=CC=C(OC)C=C1
Molecular Weight (g/mol) 184.19
Formula Weight 184.19
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation