Learn More
Tyrosine kinase inhibitor, MedChemExpress
Product Code. 16486677
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16486677 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16486677 Supplier Medchem Express Supplier No. HY10421
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
MedChemExpress Tyrosine kinase inhibitor is a potent tyrosine kinase inhibitor.
Tyrosine kinase inhibitor is a potent tyrosine kinase inhibitor.
Specifications
Chemical Name or Material Tyrosine kinase inhibitor
CAS 1021950-26-4
Color Off-White
Purity Grade Notes Research
Molecular Formula C31H31FN6O5
Quantity 5 mg
Solubility Information DMSO : ≥ 46 mg/mL (78.42 mM)
SMILES O=C(NC1=CC=C(C=C1)F)C2(CC2)C(NC3=CC=C(C=C3)OC4=C5C(NC(C(NCCN6CCOCC6)=O)=C5)=NC=C4)=O
Molecular Weight (g/mol) 586.61
Formula Weight 586.61
Percent Purity 99.96%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease