missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Type 90 Safety Storage Cabinet Q-PEGASUS-90 Winged Doors
Safe and approved storage of flammable hazardous substances in working areas
Brand: asecos™ 3000204030007
Additional Details : Weight : 436.97519kg
Alert:
For indoor use only
Description
- Safe and approved storage of flammable hazardous substances in working areas
- Robust construction and longevity: triple hinged door, safety elements assembled outside the storage compartment for increased protection against corrosion, scratch- and impact-resistant surface, easy to clean
- Effortless use: one-hand operation for double-door cabinets (AGT)
- User-friendly and safe: door open arrest system (standard), convenient and safe removal or storage of containers
- Always closed: serial automatic door closing (TSA) releases arrested doors after approx. 60 seconds; acoustic and visual signal before closing
- No unauthorised use: doors lockable with cylinder locking (integration in an existing locking system possible) and locking state indicator (red/green)
- Easy transport: integrated transport base with removable, optional base cover
- Easy alignment: adjusting aids to compensate for uneven floor
- Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system, spinning wheel in the exhaust grid as an indicator of sufficient technical ventilation
Specifications
1647 mm | |
3 x tray shelf, 1 x perforated insert (111 mm high), 1 x bottom collecting sump (sheet steel powder-coated) | |
Safe and approved storage of flammable hazardous substances in working areas | |
522 mm | |
Steel | |
615 x 1193 x 1953 mm | |
522 x 1050 x 1647 mm | |
Tray shelf | |
1953 mm | |
Q90.195.120.WDAC | |
90 mm | |
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant |
Warning Yellow doors | |
Safety storage cabinet | |
1050 mm | |
Powder-coated textured | |
1193 mm | |
483.50 kg | |
Anthracite Gray, Warning Yellow | |
615 mm | |
Wing, Auto-close | |
3 | |
2 |