missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Type 90 Combi Safety Storage Cabinets K-UB-90
Safe and approved storage of aggressive and flammable hazardous substances in working areas
Brand: asecos™ 3037700131107
Additional Details : Weight : 205.80119kg
Alert:
For indoor use only
Description
- Robust construction and longevity: body made of powder-coated sheet steel
- User-friendly and safe: door and drawer remain open in any position, interior of the cabinet completely visible — access to all containers, 93° opening of the wing door
- Safe: drawer fitted with earthing wire incl. clamp as standard, seal welded sump, drawers are self-closing in the event of fire
- No unauthorised use: door and drawer lockable with cylinder lock and locking state indicator (red/green)
- Mobile: castors with plinth (optionally available), easy moving of the cabinet
- Ventilation: integrated air ducts ready for connection (DN 50) to a technical exhaust system, extraction of both compartments through one ventilation connection
- Certificates
- Fire resistance 90 minutes
- Type tested in accordance with EN 14470-1
- EN 16121/16122 conformity
- CE compliant
Specifications
502 mm | |
Combi safety storage cabinets | |
770 mm | |
Powder-coated plain | |
1402 mm | |
196.50 kg | |
Light Gray | |
502 mm | |
Wing | |
2 | |
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, CE compliant |
1 x perforated insert, 1 x 11 liter bottom collecting sump (sheet steel powder-coated), 2 x pull-out shelf (panel melamine resin-coated/PP) | |
Safe and approved storage of aggressive and flammable hazardous substances in working areas | |
380 mm | |
Steel | |
502 x 1402 x 601 mm | |
Left: 380 x 770 x 502 mm, Right: 468 x 440 x 562 mm | |
Pull-out | |
601 mm | |
K90.060.140.050.UB.3T | |
3 |