missing translation for 'onlineSavingsMsg'
Learn More

TXNRD2 Antibody - BSA Free, Novus Biologicals™

Product Code. 18623162 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.02 mL
0.1 mL
Unit Size:
0.02mL
0.1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18623162 0.1 mL 0.1mL
18684052 0.02 mL 0.02mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18623162 Supplier Novus Biologicals Supplier No. NBP2942560.1ml

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TXNRD2 Polyclonal antibody specifically detects TXNRD2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen TXNRD2
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin
Formulation PBS (pH 7.3), 50% glycerol
Gene Alias EC 1.8.1, EC 1.8.1.9, KIAA1652, Selenoprotein Z, SelZ, thioredoxin reductase 2, thioredoxin reductase 3, thioredoxin reductase beta, Thioredoxin reductase TR3, TR, TR3, TR-BETA, TRXR2thioredoxin reductase 2, mitochondrial
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 100-310 of human TXNRD2 (NP_006431.2). AALLGGLIQDAPNYGWEVAQPVPHDWRKMAEAVQNHVKSLNWGHRVQLQDRKVKYFNIKASFVDEHTVCGVAKGGKEILLSADHIIIATGGRPRYPTHIEGALEYGITSDDIFWLKESPGKTLVVGASYVALECAGFLTGIGLDTTIMMRSIPLRGFDQQMSSMVIEHMASHGTRFLRGCAPSRVRRLPDGQLQVTWEDSTTGKEDTGTFD
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Membrane Trafficking and Chaperones
Primary or Secondary Primary
Gene ID (Entrez) 10587
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.