Learn More
TX1-85-1, MedChemExpress
Product Code. 16495917
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
Product Code. 16495917
Brand: Medchem Express HY100848
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress TX1-85-1 is an irreversible Her3 (ErbB3) inhibitor with an IC50 of 23 nM. TX1-85-1 is also the first selective Her3 ligand, which forms a covalent bond with Cys721 located in the ATP-binding site of Her3. TX1-85-1 induces partial degradation of Her3 protein and attenuates Her3-dependent signaling.
TX1-85-1 is an irreversible Her3 (ErbB3) inhibitor with an IC50 of 23 nM. TX1-85-1 is also the first selective Her3 ligand, which forms a covalent bond with Cys721 located in the ATP-binding site of Her3. TX1-85-1 induces partial degradation of Her3 protein and attenuates Her3-dependent signaling.
Specifications
Chemical Name or Material TX1-85-1
CAS 1603845-32-4
Color White
Purity Grade Notes Research
Molecular Formula C32H36N8O3
Quantity 5 mg
Solubility Information DMSO : 3.85 mg/mL (6.63 mM; Need ultrasonic)
SMILES CC(N1CCN(C2CCC(N3N=C(C4=CC(NC(C=C)=O)=C(OC5=CC=CC=C5)C=C4)C6=C3N=CN=C6N)CC2)CC1)=O
Molecular Weight (g/mol) 580.68
Formula Weight 580.68
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease