Learn More
SI Analytics™ TW Alpha Plus Sample Changer
Shop All SI Analytics Products
Click to view available options
Volume (Metric) Sample:
50 mL
Unit Size:
Each
Description
- Sample changer TW alpha plus
- Basic unit TW alpha plus with sample rack TZ 1454 for 24 samples
- Includes titration head TZ 1462, main cable, connection cable TZ 3084 and 30 beakers
- High form
Specifications
Specifications
| Type | Sample Changer |
| Includes | Titration Head TZ 1469, Main Cable, Connection Cable TZ3084, and 30 Beakers, 50mL. |
| Model | TW Alpha Plus 24 |
| Volume (Metric) Sample | 50 mL |
| Voltage | 230 V |