Learn More
Fisherbrand™ Turbo PTFE Stir Bar Retriever with Rare Earth Core
Product Code. 16245871 Shop All Fisherbrand Products
Click to view available options
Length (Metric):
350 mm
450 mm
Unit Size:
Each
Product Code. 16245871
Please to purchase this item. Need a web account? Register with us today!
Fisherbrand™ Turbo PTFE Stir Bar Retriever with Rare Earth Core are used to retrieve stir bars from mixing containers. The PTFE stir bar retriever is chemically resistant, inert, and leak-free. Uses Samarium-Cobalt magnets at its core.
  • Inert and leak-free
  • Molded in pure, FDA-approved PTFE
  • Alnico V magnets
  • Autoclavable
  • For girant stirrer bars
Specifications
Color White
Description Turbo PTFE Stir Bar Retriever with Rare Earth Core for Giant Stir Bars
For Use With (Equipment) For the removal of Giant Stir Bars from vessels of all kinds
Diameter (Metric) 18 mm
Length (Metric) 450 mm
Material PTFE with Rare Earth Magnet Core
Quantity 1