missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TULA/STS-2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-00200-50ul
This item is not returnable.
View return policy
Description
TULA/STS-2 Polyclonal antibody specifically detects TULA/STS-2 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)
Specifications
| TULA/STS-2 | |
| Polyclonal | |
| ELISA 1:5000-1:10000, Immunohistochemistry-Paraffin 1:500-1:2000 | |
| Cbl-interacting protein 4, gene similar to UBA containing SH3 domain10CLIP4STS-2T-cell ubiquitin ligand protein, STS2, Suppressor of T-cell receptor signaling 2, T-cell ubiquitin ligand, TULAubiquitin-associated and SH3 domain-containing protein A, ubiquitin associated and SH3 domain containing A | |
| Produced in rabbits immunized with purified, recombinant Human TULA/STS-2 (Accession#: AAH69511.1; Ala354-Asn623) | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 53347 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction