missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Tucatinib

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Tucatinib is a potent and selective ErbB-2 (also called HER2) kinase inhibitor (IC50 = 7 nM). It exhibits >50-fold selectivity for ErbB-2 over EGFR. In vitro, Tucatinib inhibits ErbB-2 signaling activity and cell proliferation in ErbB-2-amplified breast cancer cell lines (BT-474). In vivo, Tucatinib (25-100 mg/kg) reduces tumor volume in breast and gastric xenograft mouse models.This compound is orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | N6-(4,5-Dihydro-4,4-dimethyl-2-oxazolyl)-N4-[3-methyl-4-([1,2,4 ]triazolo[1,5-a]pyridin-7-yloxy)phenyl]-4,6-quinazolinediamine |
| CAS | 937263-43-9 |
| Quantity | 50 mg |
| Target | EGF Receptor Inhibitors |
| Molecular Formula | C26H24N8O2 |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction