missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tubulin alpha-8 Antibody (2D6), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
217.00 € - 557.00 €
Specifications
| Antigen | Tubulin alpha-8 |
|---|---|
| Clone | 2D6 |
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18156008
|
Novus Biologicals
NBP2-37535 |
0.1 mL |
557.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18624366
|
Novus Biologicals
NBP2-37535-0.025ml |
0.025 mL |
217.00 €
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tubulin alpha-8 Monoclonal specifically detects Tubulin alpha-8 in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Tubulin alpha-8 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Ascites with 0.03% Sodium Azide | |
| 51807 | |
| Purified recombinant fragment of human TUBA8 expressed in E. Coli. | |
| Primary | |
| 50 kDa |
| 2D6 | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human, Rat | |
| Alpha-tubulin 8, TUBAL2Tubulin alpha chain-like 2, tubulin alpha-8 chain, tubulin, alpha 8 | |
| TUBA8 | |
| IgG2b | |
| Unpurified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title