Learn More
Upchurch Scientific Tubing SST 1/16 inch O.D. 0.040 inch (1.02mm)I.D. 0.5m
Brand:  Upchurch Scientific 02011-40
Tubing SST 1/16 inch O.D. 0.040 inch (1.02mm)I.D. 0.5m
Product Code. 13040910
  • ONLINE EXCLUSIVE PRICE
    86.25 €
    65.89 € / Each
    Save 20.36 €
    23% Off
Only null left
Estimated Shipment: 23-01-2026