Learn More
SymLine FlexTube Screw Connection
Tubing screw connection constructed of chemically resistant PTFE.
Brand: SymLine 106591
67.77 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- PTFE construction for chemical resistance
- Secure, screw-type connection
- Suitable for FlexTube tubing
Specifications
Screw Connection | |
FlexTube Pro GL25 Tubing | |
Black |
PTFE | |
2 Pk. |